6-FAM phosphoramidite [5'-Fluorescein phosphoramidite] ≥90%
Supplier: AAT BIOQUEST
Certificates
About this item
Synonyms: 6-Fluorescein Phosphoramidite, 6-((6-(((2-Cyanoethoxy)(diisopropylamino)phosphaneyl)oxy)hexyl)carbamoyl)-3-oxo-3H-spiro[isobenzofuran-1,9'-xanthene]-3',6'-diyl bis(2,2-dimethylpropanoate), 5'-Fluorescein CE-Phosphoramidite (6-FAM)
CAS Number:204697-37-0
:
Formula:C₄₆H₅₈N₃O₁₀P
MDL Number:MFCD01940925
Molecular Weight:843.94 g/mol
ADR:5 II
:GHS07
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together