Order Entry
Belgium
ContactUsLinkComponent
TRITON™ X-100 (Polyethylene glycol tert-octylphenyl ether) 10% in water, ReadiUse™ Peroxide-and Carbonyl-Free
TRITON™ X-100 (Polyethylene glycol tert-octylphenyl ether) 10% in water, ReadiUse™ Peroxide-and Carbonyl-Free
Catalog # AATB60012
Supplier:  AAT BIOQUEST
CAS Number:  
undefined
TRITON™ X-100 (Polyethylene glycol tert-octylphenyl ether) 10% in water, ReadiUse™ Peroxide-and Carbonyl-Free
Catalog # AATB60012
Supplier:  AAT BIOQUEST
CAS Number:  

Specifications

  • Pk:
    50 mL
  • Seq:
    2370
  • Environmentally Preferable:

Specifications

About this item

Synonyms: Polyethyleneglycol-[4-(1,1,3,3-tetramethylbutyl)phenyl]-ether, (p-tert-Octylphenoxy)polyethoxyethanol, Octoxynol, Polyoxyethylene(10) Octylphenyl Ether, t-Octylphenoxypolyethoxyethanol, O-[4-(1,1,3,3-Tetramethylbutyl)phenoxy]polyethoxyethanol
 :
Storage Temperature:Ambient
CAS Number:9002-93-1
 :9036-19-5
MDL Number:MFCD00132505