Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Belgium
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
5-TAMRA cadaverine
5-TAMRA cadaverine ≥95%
5-TAMRA cadaverine ≥95%
Catalog # AATB356
Supplier:
AAT BIOQUEST
CAS Number:
Expand image
5-TAMRA cadaverine ≥95%
Catalog # AATB356
Supplier:
AAT BIOQUEST
CAS Number:
Specifications
Pk:
5 mg
Seq:
2210
Specifications
About this item
Product Details
:
:
514.62 g/mol
Formula:
C₃₀H₃₄N₄O₄
Product Details