Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Belgium
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
6-Carboxyrhodamine 6G (6-CR6G)
6-Carboxyrhodamine 6G (6-CR6G) single isomer ≥95%
6-Carboxyrhodamine 6G (6-CR6G) single isomer ≥95%
6-Carboxyrhodamine 6G (6-CR6G) single isomer ≥95%
Catalog # AATB332
Supplier:
AAT BIOQUEST
CAS Number:
Expand image
6-Carboxyrhodamine 6G (6-CR6G) single isomer ≥95%
Catalog # AATB332
Supplier:
AAT BIOQUEST
CAS Number:
Specifications
Pk:
10 mg
Seq:
2275
Specifications
About this item
Product Details
:
CAS Number:
180144-68-7
Product Details