Order Entry
Belgium
ContactUsLinkComponent
PE-iFluor™ 750 Tandem, ReadiUse™ fluorescent probe, preactivated
Irritant
PE-iFluor™ 750 Tandem, ReadiUse™ fluorescent probe, preactivated
Catalog # AATB2578
Supplier:  AAT Bioquest
CAS Number:  
undefined
Irritant
PE-iFluor™ 750 Tandem, ReadiUse™ fluorescent probe, preactivated
Catalog # AATB2578
Supplier:  AAT Bioquest
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    1 mg
  • Seq:
    2070

Specifications

About this item

 :
 :240000 g/mol
ADR:5,II
 :GHS07