R-Phycoerythrin
Supplier: AAT Bioquest
Jump to:
About this item
R-Phycoerythrin (PE) is isolated from red algae.
- CAS Number:11016-17-4
- MDL Number:MFCD00081950
- Molecular Weight:240 kDa
- ADR:5,II
3 Options Available Below
Product Details & Documents
R-Phycoerythrin (PE) is isolated from red algae.
- CAS Number:11016-17-4
- MDL Number:MFCD00081950
- Molecular Weight:240 kDa
- ADR:5,II
Product Family Options
Product Information
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



