Order Entry
Belgium
Orders LinkContactUsLinkComponent
 

Thioflavin T ≥95%

Plastic bottle, 1 g
Catalog # AATB23060
Irritant
Supplier:   AAT Bioquest
Thioflavin T ≥95%
Catalog # AATB23060
Supplier:  AAT Bioquest
CAS Number:  2390-54-7
Thioflavin T ≥95%
Thioflavin T ≥95%

Plastic bottle, 1 g

CAS Number:  2390-54-7
Price
122,00 €
Log in for availability
1 * 1 g
Product Details & Documents

Thioflavin T (ThT), also called Basic Yellow 1 or CI 49005, is a benzothiazole salt.

Synonyms:  CI 49005, Thioflavine T, Basic Yellow 1, 2-(4-(Dimethylamino)phenyl)-3,6-dimethylbenzo[d]thiazol-3-ium chloride, Setoflavine T
  • CAS Number:2390-54-7
  • Formula:C₁₇H₁₉ClN₂S
  • MDL Number:MFCD00011944
  • Molecular Weight:318.86 g/mol
  • ADR:5,II
Specifications

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    1 g

Specifications