Order Entry
Belgium
ContactUsLinkComponent
 
 
5(6)-Carboxyfluorescein diacetate NHS ester [CFSE (5(6)-CFDA SE)] ≥95%
  AATB22022
 :  
undefined
 
5(6)-Carboxyfluorescein diacetate NHS ester [CFSE (5(6)-CFDA SE)] ≥95%
  AATB22022
 :  

 

  • Pack type:
    Plastic bottle
  • Pk:
    25 mg
  • Seq:
    2315

 

 

 :
 :557.467 g/mol
 :5,II
 :GHS07
 :150347-59-4
 :MFCD31812843
 :C₂₉H₁₉NO₁₁
 :MFCD00037466