Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Belgium
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Dextran
Dextran, M.W. 150,000 FITC labelled
Dextran, M.W. 150,000 FITC labelled
Dextran, M.W. 150,000 FITC labelled
Catalog # AATB21704
Supplier:
AAT BIOQUEST
CAS Number:
Expand image
Dextran, M.W. 150,000 FITC labelled
Catalog # AATB21704
Supplier:
AAT BIOQUEST
CAS Number:
Specifications
Pack type:
Vial
Pk:
25 mg
Seq:
2315
Specifications
About this item
Product Details
:
CAS Number:
9004-54-0
Formula:
(C₆H₁₀O₅)n
MDL Number:
MFCD00130935
Product Details