Order Entry
Belgium
ContactUsLinkComponent
DiBAC₄(3) for membrane potential measurement
Irritant
DiBAC₄(3) for membrane potential measurement
Catalog # AATB21411
Supplier:  AAT Bioquest
CAS Number:  
undefined
Irritant
DiBAC₄(3) for membrane potential measurement
Catalog # AATB21411
Supplier:  AAT Bioquest
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    25 mg
  • Seq:
    2315

Specifications

About this item

 :
 :516.63 g/mol
ADR:5,II
 :GHS07
CAS Number:70363-83-6
Formula:C₂₇H₄₀N₄O₆
MDL Number:MFCD07437857