Order Entry
Belgium
Orders LinkContactUsLinkComponent
Zinquin

Zinquin

Plastic bottle, 5 mg
Catalog # AATB21260
Irritant
Supplier:   AAT Bioquest
Zinquin
Catalog # AATB21260
Supplier:  AAT Bioquest
CAS Number:  151606-29-0
Zinquin
Zinquin

Plastic bottle, 5 mg

CAS Number:  151606-29-0
Price
156,00 €
Log in for availability
1 * 5 mg
Product Details & Documents

Zinquin is a zinc-sensitive, cell-impermeable fluorescent probe.

Synonyms:  Zinquin A
  • CAS Number:151606-29-0
  • Formula:C₁₉H₁₈N₂O₅S
  • MDL Number:MFCD03452711
  • Molecular Weight:386.42 g/mol
  • ADR:5,II
Specifications

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    5 mg

Specifications