Order Entry
Belgium
ContactUsLinkComponent
Dextran, M.W. 10,000 Cal-590™ labelled
Dextran, M.W. 10,000 Cal-590™ labelled
Catalog # AATB20509
Supplier:  AAT BIOQUEST
CAS Number:  
undefined
Dextran, M.W. 10,000 Cal-590™ labelled
Catalog # AATB20509
Supplier:  AAT BIOQUEST
CAS Number:  

Specifications

  • Pack type:
    Vial
  • Pk:
    1 mg
  • Seq:
    2070

Specifications

About this item

 :
CAS Number:9004-54-0
Formula:(C₆H₁₀O₅)n
MDL Number:MFCD00130935