Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Belgium
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
OG488 BAPTA-1 AM
OG488 BAPTA-1 AM Cell permeant [equivalent to Oregon Green® 488 BAPTA-1, AM]
OG488 BAPTA-1 AM Cell permeant [equivalent to Oregon Green® 488 BAPTA-1, AM]
AATB20507
:
AAT Bioquest
:
OG488 BAPTA-1 AM Cell permeant [equivalent to Oregon Green® 488 BAPTA-1, AM]
AATB20507
:
AAT Bioquest
:
Pack type:
Plastic bottle
Pk:
500 µG
Seq:
1418
:
:
1258.07 g/mol
:
5,II
:
GHS07
:
244167-57-5
:
C59H53F2N3O26