Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Belgium
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Cy7NS acid
Cy7NS acid ≥90%
Cy7NS acid ≥90%
Cy7NS acid ≥90%
Catalog # AATB197
Supplier:
AAT BIOQUEST
CAS Number:
Expand image
Cy7NS acid ≥90%
Catalog # AATB197
Supplier:
AAT BIOQUEST
CAS Number:
Specifications
Pk:
100 mg
Seq:
2440
Specifications
About this item
Product Details
:
:
508.69 g/mol
Product Details