
Aminoallyl UTP sodium salt, (AA-UTP) 4 mM in Tris/EDTA (TE) buffer
Catalog # AATB17021
Supplier: AAT Bioquest
CAS Number:
Specifications
- Pack type:Plastic bottle
- Pk:250 µl
- Seq:1370
Specifications
About this item
:
:605.16 g/mol
ADR:5,II
:GHS07
CAS Number:75221-88-4