Order Entry
Belgium
Orders LinkContactUsLinkComponent
Ac-Pro-Ala-Leu-AMC fluorogenic substrate

Ac-Pro-Ala-Leu-AMC fluorogenic substrate

Plastic bottle, 5 mg
Catalog # AATB13479
Irritant
Supplier:   AAT Bioquest
Ac-Pro-Ala-Leu-AMC fluorogenic substrate
Catalog # AATB13479
Supplier:  AAT Bioquest
CAS Number:  1431362-79-6
Ac-Pro-Ala-Leu-AMC fluorogenic substrate
Ac-Pro-Ala-Leu-AMC fluorogenic substrate

Plastic bottle, 5 mg

CAS Number:  1431362-79-6
Price
156,00 €
Log in for availability
1 * 5 mg
Product Details & Documents

The weakly fluorescent AMC substrates generate the bright blue fluorescent AMC product that has Ex/Em = 351/430 nm, and can be easily detected with a DAPI filter set.

  • High fluorescence
  • Rapid activation
  • CAS Number:1431362-79-6
  • Molecular Weight:498.57 g/mol
  • ADR:5,II
Specifications

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    5 mg

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.