Order Entry
Belgium
ContactUsLinkComponent
D-Luciferin 6′-O-phosphate trisodium salt ≥90%
Irritant
D-Luciferin 6′-O-phosphate trisodium salt ≥90%
Catalog # AATB12512
Supplier:  AAT Bioquest
CAS Number:  
undefined
Irritant
D-Luciferin 6′-O-phosphate trisodium salt ≥90%
Catalog # AATB12512
Supplier:  AAT Bioquest
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    1 mg
  • Seq:
    2070

Specifications

About this item

 :
ADR:5,II
 :426.25 g/mol
 :GHS07
CAS Number:145613-12-3
Formula:C₁₁H₆N₂Na₃O₆PS₂
MDL Number:MFCD00214322