Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Belgium
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Glucose-UDP-(PEG)6-Fluorescein
Glucose-UDP-(PEG)6-Fluorescein for conjugation
Glucose-UDP-(PEG)6-Fluorescein for conjugation
Irritant
Glucose-UDP-(PEG)6-Fluorescein for conjugation
Catalog # AATB11706
Supplier:
AAT Bioquest
CAS Number:
Expand image
Irritant
Glucose-UDP-(PEG)6-Fluorescein for conjugation
Catalog # AATB11706
Supplier:
AAT Bioquest
CAS Number:
Specifications
Pack type:
Plastic bottle
Pk:
100 µG
Seq:
1286
Specifications
About this item
Product Details
:
ADR:
5,II
:
GHS07
Product Details