Order Entry
Belgium
Orders LinkContactUsLinkComponent
3,3',5,5'-Tetramethylbenzidine (TMB) substrate, ReadiUse™ (colorimetric)

3,3',5,5'-Tetramethylbenzidine (TMB) substrate, ReadiUse™ (colorimetric)

Irritant
Supplier:   AAT Bioquest
3,3',5,5'-Tetramethylbenzidine (TMB) substrate, ReadiUse™ (colorimetric)
Supplier:  AAT Bioquest
Certificates
3,3',5,5'-Tetramethylbenzidine (TMB) substrate, ReadiUse™ (colorimetric)
3,3',5,5'-Tetramethylbenzidine (TMB) substrate, ReadiUse™ (colorimetric)

2 Options Available Below

Product Details & Documents

Horseradish peroxidase (HRP) and HRP conjugates facilitate the ABTS oxidation in the presence of hydrogen peroxide, turning ABTS into its blue-green oxidized product.

  • Easy colorimetric detection
  • Easily measured via spectrophotometer
Synonyms:  TMB, 3,3',5,5'-Tetramethyl-[1,1'-biphenyl]-4,4'-diamine
  • CAS Number:54827-17-7
  • Formula:C₁₆H₂₀N₂
  • MDL Number:MFCD00007748
  • Molecular Weight:240.35 g/mol
  • ADR:5,II
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Plastic bottle
    100 mL
    Log in for availability
    Log in for availability
    1 * 100 mL
    1 * 100 mL - 122,00 €
    122,00 €
    1 * 100 mL
       122,00 €

  • Plastic bottle
    1 L
    Log in for availability
    Log in for availability
    1 * 1 L
    1 * 1 L - 785,00 €
    785,00 €
    1 * 1 L
       785,00 €

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.