Order Entry
Belgium
ContactUsLinkComponent
EX-LINE GAPEX Hazardous Material Work Station
EX-LINE GAPEX Hazardous Material Work Station
Catalog # MSGA40585901
Supplier:  asecos
EX-LINE GAPEX Hazardous Material Work Station
Catalog # MSGA40585901
Supplier:  asecos

Specifications

  • Weight:
    131,70 kg
  • W×D×H ext.:
    900×750×1400 mm mm
  • Description:
    Hazardous material work station, EX-LINE GAPEX model GAP.140.090.EX
  • Pk:
    1
  • W×D×H int.:
    865×650×1110 mm mm

Specifications

About this item

Hazardous material work station, EX-LINE GAPEX model with robust construction made of chemically resistant, anodised aluminium profiles.

  • Required occupational exposure limits are maintained
  • used for testing of the air equipment according to EN 14175 Part 3 (5.4.4)
  • Maximum user safety
  • Transparent side panes made from 5 mm (toughened safety glass)
  • Highly effective capturing of hazardous materials inside the work station through special fresh air curtain technology
  • Prepared for connection to the on-site exhaust air system

Standard equipment consists of:
Control box with complete monitoring electronics incl. potential-free switching contact, e.g. for controlling on-site exhaust air system (must be installed outside the Ex zone) - Cable length 5 m
Transparent side panes made from 5 mm (toughened safety glass)
Fold-away deflector
Melamine resin coated rear wall
Working surface made from stainless steel (V2A/1.4301) as seal welded sump
Support frame for standing work
Earthing link, mounted in the profile
EX-LED lamp 6500 K

Certifications: Electrical equipment approved in accordance with 2014/34/EU.