Orders LinkContactUsLinkComponent
Digital Microscope
Digital Microscope
10.5×3.2 cm, Digital, 5 Megapixel (2592×1944), Standard
Catalog # IDCPWF7915MZT
Digital Microscope
Catalog # IDCPWF7915MZT
Digital Microscope
10.5×3.2 cm, Digital, 5 Megapixel (2592×1944), Standard
Price
1 710,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
With the use of the latest, cutting-edge optics, a brand new 5 megapixel sensor and several special features, it has a marvel of technology and the best choice for the high-demanding professional. It offers superb image quality and color reproduction in a robust, compact and appealing housing.
  • Wireless
  • Extended depth of field (EDOF)
  • Extended dynamic range (EDR)
  • Flexible LED control (FLC)
  • Automatic magnification reading (AMR)
With the Extended Dynamic Range (EDR) feature, the details of darker or brighter areas within the object can be revealed by stacking images at different exposure levels. The Extended Depth of Field (EDOF) feature automatically stacks images at different focus level to improve the depth of field on rough or uneven surfaces. With the built-in Automatic Magnification Reading (AMR), measurements can be performed easily and quickly. Because of the built-in polarization filter this model is ideal when working with shiny or reflective objects such as metal, plastic, glass, jewelry, electronics, etc.
Provides 2 years warranty.
Packaging: Microscope, carry pouch, software CD, calibration target, user manual, N3C-O- open cap, N3C-C closed cap, N3C-D- diffuser cap, N3C-E extension cap, N3C-L- Long cap, N3C-S- side light cap.
Specifications
  • Magnification:
    10× ~ 220×
  • Resolution:
    5 Megapixel (2592×1944)
  • Description:
    10.5×3.2 cm
  • Working distance:
    Standard
  • Type:
    Digital
  • Cat. No.:
    IDCPWF7915MZT
  • Software:
    DinoCapture 2.0 (Windows), DinoXcope (Mac
    OS)
  • Weight:
    137 g
  • Sensor:
    CMOS