Order Entry
Belgium
Orders LinkContactUsLinkComponent
Digital Microscope

Digital Microscope

130×30 mm, Digital, 8 Megapixel (3840×2160), 142,6 ~ 11,5 mm
Catalog # IDCPAM8117MZT
Digital Microscope
Catalog # IDCPAM8117MZT
Digital Microscope

130×30 mm, Digital, 8 Megapixel (3840×2160), 142,6 ~ 11,5 mm

Price
1 080,00 €
Log in for availability
1 * 1 ST
Product Details & Documents

Taking image quality to the next level, the 8-megapixel, this digital microscope delivers high resolution and color fidelity for a natural-looking and detailed imaging experience.

  • High optical resolution
  • Enhanced flexible LED Control (eFLC)
  • Adjustable polariser
  • Robust metal housing
  • 10× ~ 220× magnification (Edge shading occurs below 25× when previewing in 16:9 windows.) french, spanish and more

Includes a polariser for reducing unwanted reflections as well as eFLC for maximising lighting adjustability.

2 years warranty.

Packaging: N3C-C (close cap) N3C-D (diffuser cap) N3CD2 (opal diffuser cap) N3C-E (extended open cap) N3C-L (Long Cap) N3C-O (open cap) N3CS (sidelight cap).

Specifications
  • Magnification:
    10× ~ 220×
  • Resolution:
    8 Megapixel (3840×2160)
  • Description:
    130×30 mm
  • Working distance:
    142,6 ~ 11,5 mm
  • Type:
    Digital
  • Cat. No.:
    IDCPAM8117MZT
  • Frame rate:
    10 fps at 8,0 MP, MJPEG 25 fps at 2,0 MP, MJPEG
  • Software:
    Windows 10/11
  • Weight:
    120 g
  • Sensor:
    CMOS
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.