Order Entry
Belgium
Orders LinkContactUsLinkComponent
Separating funnel holder

Separating funnel holder

for 1 separating funnel up from 75 mm Ø
Catalog # 241-0345
Supplier:   Brand
Separating funnel holder
Catalog # 241-0345
Supplier:  Brand
Separating funnel holder
Separating funnel holder

for 1 separating funnel up from 75 mm Ø

Price
12,50 €
Log in for availability
1 * 1 ST
Product Details & Documents

PP, white.

For conical separating funnels up to 1000 ml. Easely mounted on support rods with an outer Ø 8 to 14 mm with a locking nut.

Specifications
  • Description:
    for 1 separating funnel up from 75 mm Ø
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.