Order Entry
Belgium
ContactUsLinkComponent
(±)-2-Methyltetrahydrofuran ≥99.0% stabilised, GPR RECTAPUR® for synthesis
Flammable
(±)-2-Methyltetrahydrofuran ≥99.0% stabilised, GPR RECTAPUR® for synthesis
Supplier:  VWR Chemicals
Certificates
undefined
Flammable
(±)-2-Methyltetrahydrofuran ≥99.0% stabilised, GPR RECTAPUR® for synthesis
Supplier:  VWR Chemicals
Certificates

Specification Test Results

  • Appearance:
    Clear colourless liquid
  • Assay:
    Min. 99.0 %
  • IR Spectrum:
    Passes test
  • Water:
    Max. 0.03 %

Specification Test Results

Frequently Bought Together