Orders LinkContactUsLinkComponent
GAP Hazardous Material Work Stations
Supplier:   asecos
GAP Hazardous Material Work Stations
Supplier:  asecos
Certificates
GAP Hazardous Material Work Stations
13 Options Available Below
Product Details & Documents
Hazardous material work station, GAP model with robust construction made of chemically resistant, anodised aluminium profiles.
  • Required occupational exposure limits are maintained
  • Avoidance of dangerous explosive atmosphere
  • Used for testing of the air equipment according to EN 14175 Part 3 (5.4.4)
  • Maximum user safety
  • Transparent side panes made from 5 mm (toughened safety glass)
  • Highly effective capturing of hazardous materials inside the work station through special fresh air curtain technology
  • Prepared for connection to the on-site exhaust air system and for electrical supply (230 V/50 Hz)
  • For mounting on existing working surfaces
Standard equipment consists of:
Melamine resin coated rear wall
Exhaust and fresh air monitoring with visual and audible alarm
Interior lighting aranged anti-glare (LED 4000 K neutral white 500 lux on the work surface)
Transparent side walls
Opaque, fold-away deflector
Potential-free switching contact 4-pole (max.400 V/25 A)
Power cord 3 m
Product Family Options
Product Information
  • W×D×H ext.
    W×D×H int.
    Weight
    Availability
    Price
  • MSGA40563901
    Hazardous material work station GAP.110.090.060
    Hazardous material work station GAP.110.090.060
    900×600×1100 mm
    865×500×855 mm
    92,20 kg
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 6 670,00 €
    6 670,00 €
    1 * 1 ST
       6 670,00 €
  • MSGA40581901
    Hazardous material work station GAP.140.090.060
    Hazardous material work station GAP.140.090.060
    900×600×1400 mm
    865×500×1105 mm
    102,20 kg
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 6 860,00 €
    6 860,00 €
    1 * 1 ST
       6 860,00 €
  • MSGA40564901
    Hazardous material work station GAP.110.090
    Hazardous material work station GAP.110.090
    900×750×1100 mm
    865×650×855 mm
    103,20 kg
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 6 640,00 €
    6 640,00 €
    1 * 1 ST
       6 640,00 €
  • MSGA40582901
    Hazardous material work station GAP.140.090
    Hazardous material work station GAP.140.090
    900×750×1400 mm
    865×650×1105 mm
    113,20 kg
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 6 770,00 €
    6 770,00 €
    1 * 1 ST
       6 770,00 €
  • MSGA40566901
    Hazardous material work station GAP.110.120.060
    Hazardous material work station GAP.110.120.060
    1200×600×1100 mm
    1165×500×855 mm
    105,20 kg
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 6 950,00 €
    6 950,00 €
    1 * 1 ST
       6 950,00 €
  • MSGA40586901
    Hazardous material work station GAP.140.120.060
    Hazardous material work station GAP.140.120.060
    1200×600×1400 mm
    1165×500×1105 mm
    80,70 kg
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 7 130,00 €
    7 130,00 €
    1 * 1 ST
       7 130,00 €
  • MSGA40567901
    Hazardous material work station GAP.110.120
    Hazardous material work station GAP.110.120
    1200×750×1100 mm
    1165×650×855 mm
    116,70 kg
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 6 900,00 €
    6 900,00 €
    1 * 1 ST
       6 900,00 €
  • MSGA40587901
    Hazardous material work station GAP.140.120
    Hazardous material work station GAP.140.120
    1200×750×1400 mm
    1165×650×1105 mm
    127,70 kg
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 7 020,00 €
    7 020,00 €
    1 * 1 ST
       7 020,00 €
  • MSGA40569901
    Hazardous material work station GAP.110.150.060
    Hazardous material work station GAP.110.150.060
    1500×600×1100 mm
    1465×500×855 mm
    120,80 kg
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 7 780,00 €
    7 780,00 €
    1 * 1 ST
       7 780,00 €
  • MSGA40570901
    Hazardous material work station GAP.110.150
    Hazardous material work station GAP.110.150
    1500×750×1100 mm
    1465×650×855 mm
    131,80 kg
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 7 810,00 €
    7 810,00 €
    1 * 1 ST
       7 810,00 €
  • MSGA40572901
    Hazardous material work station GAP.110.180.060
    Hazardous material work station GAP.110.180.060
    1800×600×1100 mm
    1765×500×855 mm
    135,30 kg
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 8 160,00 €
    8 160,00 €
    1 * 1 ST
       8 160,00 €
  • MSGA40573901
    Hazardous material work station GAP.110.180
    Hazardous material work station GAP.110.180
    1800×750×1100 mm
    1765×650×855 mm
    152,30 kg
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 8 240,00 €
    8 240,00 €
    1 * 1 ST
       8 240,00 €
  • MSGA41044901
    Hazardous material work station GAP.140.150.060
    Hazardous material work station GAP.140.150.060
    1500×600×1400 mm
    1465×500×1110 mm
    90,80 kg
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 8 360,00 €
    8 360,00 €
    1 * 1 ST
       8 360,00 €