Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Belgium
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Transfection Reagents
BaculoFECTIN II Transfection Reagent
BaculoFECTIN II Transfection Reagent
AVIVGWB-300101
:
Aviva Systems Biology
:
BaculoFECTIN II Transfection Reagent
AVIVGWB-300101
:
Aviva Systems Biology
:
Pk:
150 µl
Description:
BaculoFECTIN II transfection reagent
Storage temperature:
Store tightly capped at –20 ℃.