Order Entry
Belgium
ContactUsLinkComponent
Gasoline Calibration Standard (2 Compound Mix) in Petroleum Ether
Gasoline Calibration Standard (2 Compound Mix) in Petroleum Ether
Catalog # REGEREGASCAL-8-250
Supplier:  REAGECON
CAS Number:  
Gasoline Calibration Standard (2 Compound Mix) in Petroleum Ether
Catalog # REGEREGASCAL-8-250
Supplier:  REAGECON
CAS Number:  

Specifications

  • Concentration:
    Concentration of DIPE is 20%, Petroleum ether is 66% and tert-butanol is 14%.
  • Pack type:
    Glass bottle, amber, Winchester
  • Pk:
    1
  • Standards:
    Gasoline calibration
  • Description:
    Gasoline calibration standard in petroleum ether
  • UN No.:
    1993
  • Physical form:
    Liquid

Specifications

About this item

Gasoline calibration standard (two compound mix) in petroleum ether is a ready to use standard used in fuel analysis as either a calibrant or analytical control solution. This standard is used in testing for a range of aromatic and oxygenate compounds in gasoline in accordance with the ASTM D5845, using infrared (IR) instrumentation. The product can also be used to validate appropriate test methods or qualify an instrument in a regulated industry. The product is QC tested on a high performance GC-MS.

  • Multi compound mix analyte
  • Mixed concentrations
  • Weight: 0,57 kg
  • Packing group II with class three
Certifications: Reagecon holds ISO 17025 accreditation (A2LA Ref: 6739.02) for calibration of laboratory balances.