ContactUsLinkComponent
Adapters, Cone without Stopcock
Adapters, Cone without Stopcock
Catalog # WITG1.681.029
Supplier:  witeg Labortechnik
Adapters, Cone without Stopcock
Catalog # WITG1.681.029
Supplier:  witeg Labortechnik
Specifications
  • Cone size:
    29/32 NS
  • Seq:
    05
  • Connector:
    Tubing
  • Pk:
    1
Specifications
About this item
Borosilicate glass 3.3.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion
Suction tube, with cone and without stopcock, bent.