Order Entry
Belgium
ContactUsLinkComponent
Human IgM Isotype Control (Biotin)
Human IgM Isotype Control (Biotin)
Catalog # NOVUNBP1-96989
Supplier:  Novus Biologicals
Human IgM Isotype Control (Biotin)
Catalog # NOVUNBP1-96989
Supplier:  Novus Biologicals

Specifications

  • Antibody type:
    Control
  • Antigen name:
    AGM1
  • Antigen symbol:
    IgM
  • Conjugation:
    Biotin
  • Host:
    Human
  • Isotype:
    IgM
  • Reactivity:
    Human
  • Gene ID:
    3507
  • Antigen synonyms:
    DKFZp686I15212|VH|DKFZp686I15196|MGC104996|immunoglobulin heavy constant mu|FLJ00385|MGC52291|MU
  • Storage buffer:
    0,02 M Potassium Phosphate, 0,15 M Sodium Chloride, pH 7,2. Stabilizer: 10 mg/ml BSA Immunoglobulin and Protease free.
  • Storage temperature:
    Store at 4 °C short term. Aliquot and store at −20 °C long term. Avoid freeze-thaw cycles.
  • Purification:
    Ion exchange chromatography
  • Pk:
    1 mg

Specifications

About this item

Human IgM Protein [Biotin]. Tested Reactivity: Human.

Type: Control
Antigen: AGM1
Clonality:
Clone:
Conjugation: Biotin
Epitope:
Host: Human
Isotype: IgM
Reactivity: Human