ContactUsLinkComponent
Syringe Valve Assembly Kit, 2 ml, SOCOREX® DOSYS™
Syringe Valve Assembly Kit, 2 ml, SOCOREX® DOSYS™
Catalog # WHEA851426
Supplier:  DWK Life Sciences
undefined
Syringe Valve Assembly Kit, 2 ml, SOCOREX® DOSYS™
Catalog # WHEA851426
Supplier:  DWK Life Sciences
Specifications
  • Description:
    Valve assembly kit for 2 ml self refilling syringes
  • Seq:
    03
  • Pk:
    1
Specifications
About this item
Kit includes 1 spring, 2 balls, 2 washers for valve, and barrel washer.