Orders LinkContactUsLinkComponent
SamplePro Needle Mounting Adapter
SamplePro Needle mounting adapter, H160071
H160072
H160073
Catalog # BRUKH160098
Supplier:   BRUKER DALTRONIK
SamplePro Needle Mounting Adapter
Catalog # BRUKH160098
Supplier:  BRUKER DALTRONIK
SamplePro Needle Mounting Adapter
SamplePro Needle mounting adapter, H160071
H160072
H160073
Price
206,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
SPT NADELINSERT KPL is required to mount new needle version in the SamplePro Tube liquid Handler.
Suitable for needles H160071, H160072, H160073. One insert per SamplePro Tube required.
Specifications
  • For:
    H160071
    H160072
    H160073
  • Description:
    SamplePro Needle mounting adapter