Order Entry
Belgium
ContactUsLinkComponent
Supelco® BIOshell™ IgG 1000 Å Diphenyl HPLC Columns
Supelco® BIOshell™ IgG 1000 Å Diphenyl HPLC Columns
Supplier:  Merck
Certificates
Supelco® BIOshell™ IgG 1000 Å Diphenyl HPLC Columns
Supplier:  Merck

Specifications

  • Catalog No:
  • 577432-U
  • 577433-U
  • 577430-U
  • 577431-U
  • 577428-U
  • 577429-U
  • 577451-U
  • 577450-U
  • 577440-U
  • 577439-U
  • 577438-U
  • 577437-U
  • 577427-U
  • 577425-U
  • 577426-U
  • 577423-U
  • 577424-U
  • 577421-U
  • 577422-U
  • 577419-U
  • 577420-U
  • 577434-U
  • 577436-U
  • 577435-U
  • Application:
  • For separating high molecular weight compounds such as proteins; mAbs; ADCs; Biosimilars; H/D Exchange; mAb Fragments
  • For separating high molecular weight compounds such as proteins; mAbs; ADCs; Biosimilars; H/D Exchange; mAb Fragments
  • For separating high molecular weight compounds such as proteins; mAbs; ADCs; Biosimilars; H/D Exchange; mAb Fragments
  • For separating high molecular weight compounds such as proteins; mAbs; ADCs; Biosimilars; H/D Exchange; mAb Fragments
  • For separating high molecular weight compounds such as proteins; mAbs; ADCs; Biosimilars; H/D Exchange; mAb Fragments
  • For separating high molecular weight compounds such as proteins; mAbs; ADCs; Biosimilars; H/D Exchange; mAb Fragments
  • For separating high molecular weight compounds such as proteins; mAbs; ADCs; Biosimilars; H/D Exchange; mAb Fragments
  • For separating high molecular weight compounds such as proteins; mAbs; ADCs; Biosimilars; H/D Exchange; mAb Fragments
  • For separating high molecular weight compounds such as proteins; mAbs; ADCs; Biosimilars; H/D Exchange; mAb Fragments
  • For separating high molecular weight compounds such as proteins; mAbs; ADCs; Biosimilars; H/D Exchange; mAb Fragments
  • For separating high molecular weight compounds such as proteins; mAbs; ADCs; Biosimilars; H/D Exchange; mAb Fragments
  • For separating high molecular weight compounds such as proteins; mAbs; ADCs; Biosimilars; H/D Exchange; mAb Fragments
  • For separating high molecular weight compounds such as proteins; mAbs; ADCs; Biosimilars; H/D Exchange; mAb Fragments
  • For separating high molecular weight compounds such as proteins; mAbs; ADCs; Biosimilars; H/D Exchange; mAb Fragments
  • For separating high molecular weight compounds such as proteins; mAbs; ADCs; Biosimilars; H/D Exchange; mAb Fragments
  • For separating high molecular weight compounds such as proteins; mAbs; ADCs; Biosimilars; H/D Exchange; mAb Fragments
  • For separating high molecular weight compounds such as proteins; mAbs; ADCs; Biosimilars; H/D Exchange; mAb Fragments
  • For separating high molecular weight compounds such as proteins; mAbs; ADCs; Biosimilars; H/D Exchange; mAb Fragments
  • For separating high molecular weight compounds such as proteins; mAbs; ADCs; Biosimilars; H/D Exchange; mAb Fragments
  • For separating high molecular weight compounds such as proteins; mAbs; ADCs; Biosimilars; H/D Exchange; mAb Fragments
  • For separating high molecular weight compounds such as proteins; mAbs; ADCs; Biosimilars; H/D Exchange; mAb Fragments
  • For separating high molecular weight compounds such as proteins; mAbs; ADCs; Biosimilars; H/D Exchange; mAb Fragments
  • For separating high molecular weight compounds such as proteins; mAbs; ADCs; Biosimilars; H/D Exchange; mAb Fragments
  • For separating high molecular weight compounds such as proteins; mAbs; ADCs; Biosimilars; H/D Exchange; mAb Fragments
  • Carbon load:
  • 1,00%
  • 1,00%
  • 1,00%
  • 1,00%
  • 1,00%
  • 1,00%
  • 1,00%
  • 1,00%
  • 1,00%
  • 1,00%
  • 1,00%
  • 1,00%
  • 1,00%
  • 1,00%
  • 1,00%
  • 1,00%
  • 1,00%
  • 1,00%
  • 1,00%
  • 1,00%
  • 1,00%
  • 1,00%
  • 1,00%
  • 1,00%
  • Endcapped:
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Max. pressure:
  • 600 bar
  • 600 bar
  • 600 bar
  • 600 bar
  • 600 bar
  • 600 bar
  • 600 bar
  • 600 bar
  • 600 bar
  • 600 bar
  • 600 bar
  • 600 bar
  • 600 bar
  • 600 bar
  • 600 bar
  • 600 bar
  • 600 bar
  • 600 bar
  • 600 bar
  • 600 bar
  • 600 bar
  • 600 bar
  • 600 bar
  • 600 bar
  • pH range:
  • 2 - 9
  • 2 - 9
  • 2 - 9
  • 2 - 9
  • 2 - 9
  • 2 - 9
  • 2 - 9
  • 2 - 9
  • 2 - 9
  • 2 - 9
  • 2 - 9
  • 2 - 9
  • 2 - 9
  • 2 - 9
  • 2 - 9
  • 2 - 9
  • 2 - 9
  • 2 - 9
  • 2 - 9
  • 2 - 9
  • 2 - 9
  • 2 - 9
  • 2 - 9
  • 2 - 9
  • Surface area:
  • 22 m²/g
  • 22 m²/g
  • 22 m²/g
  • 22 m²/g
  • 22 m²/g
  • 22 m²/g
  • 22 m²/g
  • 22 m²/g
  • 22 m²/g
  • 22 m²/g
  • 22 m²/g
  • 22 m²/g
  • 22 m²/g
  • 22 m²/g
  • 22 m²/g
  • 22 m²/g
  • 22 m²/g
  • 22 m²/g
  • 22 m²/g
  • 22 m²/g
  • 22 m²/g
  • 22 m²/g
  • 22 m²/g
  • 22 m²/g
  • Temperature limit:
  • 90 °C
  • 90 °C
  • 90 °C
  • 90 °C
  • 90 °C
  • 90 °C
  • 90 °C
  • 90 °C
  • 90 °C
  • 90 °C
  • 90 °C
  • 90 °C
  • 90 °C
  • 90 °C
  • 90 °C
  • 90 °C
  • 90 °C
  • 90 °C
  • 90 °C
  • 90 °C
  • 90 °C
  • 90 °C
  • 90 °C
  • 90 °C
  • USP code:
  • L11
  • L11
  • L11
  • L11
  • L11
  • L11
  • L11
  • L11
  • L11
  • L11
  • L11
  • L11
  • L11
  • L11
  • L11
  • L11
  • L11
  • L11
  • L11
  • L11
  • L11
  • L11
  • L11
  • L11

Specifications

Guard Cartridge Holder

Product Information