Order Entry
Belgium
ContactUsLinkComponent
2-Propanol standard for the analysis of tobacco
Flammable
Irritant
2-Propanol standard for the analysis of tobacco
Catalog # 6685.4000
Supplier:  VWR Chemicals
CAS Number:  
undefined
Flammable
Irritant
2-Propanol standard for the analysis of tobacco
Catalog # 6685.4000
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    4 L
  • Seq:
    5162

Specifications

Specification Test Results

  • Assay (2-Propanol) (raw material):
    ≥ 99.8 %
  • Assay (Ethanol) (raw material):
    ≥ 99.2 %
  • Assay (n-Heptadecane) (raw material):
    ≥ 99.0 %
  • Ethanol:
    2 g/l
  • n-Heptadecane:
    0.3 g/l
  • Water:
    ≤ 0.1 %

Specification Test Results

About this item

 :
 :60.1 g/mol
Boiling Pt:82 °C (1013 hPa)
Melting Pt:–89 °C
Density:0.786 g/cm³ (20 °C)
Flash Pt:12 °C
Storage Temperature:Ambient
EINECS:200-661-7
UN:1219
ADR:3,II
 :GHS02
 :GHS07
CAS Number:67-63-0
Formula:(CH₃)₂CHOH
MDL Number:MFCD00011674