Orders LinkContactUsLinkComponent
SGE OptChem™ ConnecTite Inlet Liners
SGE OptChem™ ConnecTite Inlet Liners
SGE OptChem™ ConnecTite Inlet Liners
The SGE inlet liner range aims to make it simple for all gas chromatographers to select the correct liner.
  • Easy to choose - colour coded by geometry to simplify your selection
  • Easy to use - contains O-rings so ready to go
  • Confidence in your analysis - certified deactivation
  • Robust and durable for longer life, less maintenance downtime and reduced maintenance costs
  • Color coded orange
  • Analytical formatted
ConnecTite liners facilitate maximum transfer of sample to the GC column and inhibit sample degradation by reducing contact with hot metal components inside the inlet.A hole in the liner body maintains gas flows in systems equipped with electronic pressure control. ConnecTite liners with a hole near the bottom are ideal for analyses where early eluting compounds may be affected by a tailing solvent peak.
ConnecTite liners with a hole at the top improve analyses of aqueous samples. ConnecTite liners with a hole at the top are also ideal for analyses where compounds of interest elute away from the solvent peak.
ConnecTite liners are recommended for the following sample or injection modes: Trace level analysis, direct injection, active compounds, aqueous samples.
 : ConnecTite available for Agilent, PerkinElmer, Shimadzu 2010, Thermo Scientific, and Bruker/Varian 1075/1077 injectors.
Product Family Options
  • Description
    Ext.Ø
    Ø int.
    Length
  • SGE OptChem ConnecTite inlet liners with pre-fitted CRS ONE O-ring
    6,3 mm
    4 mm
    78,5 mm
    1 * 1 ST
    1 * 1 ST - 136,00 €
    136,00 €
    1 * 1 ST
       136,00 €
  • SGE OptChem ConnecTite inlet liners with top hole and pre-fitted CRS ONE O-ring
    6,3 mm
    4 mm
    78,5
    1 * 1 ST
    1 * 1 ST - 136,00 €
    136,00 €
    1 * 1 ST
       136,00 €
  • SGE OptChem ConnecTite inlet liner with bottom hole and pre-fitted CRS ONE O-ring
    6,3 mm
    4 mm
    78,5 mm
    1 * 1 ST
    1 * 1 ST - 136,00 €
    136,00 €
    1 * 1 ST
       136,00 €
  • SGE OptChem ConnecTite inlet liners with pre-fitted CRS ONE O-ring
    6,3 mm
    4 mm
    78,5 mm
    1 * 5 ST
    1 * 5 ST - 491,00 €
    491,00 €
    1 * 5 ST
       491,00 €
  • SGE OptChem ConnecTite inlet liners with top hole and pre-fitted CRS ONE O-ring
    6,3 mm
    4 mm
    78,5 mm
    1 * 5 ST
    1 * 5 ST - 491,00 €
    491,00 €
    1 * 5 ST
       491,00 €
  • SGE OptChem ConnecTite inlet liner with bottom hole and pre-fitted CRS ONE O-ring
    6,3 mm
    4 mm
    78,5 mm
    1 * 5 ST
    1 * 5 ST - 491,00 €
    491,00 €
    1 * 5 ST
       491,00 €
CRS O-Rings
  •  
    Material
    Ø
    Description
    For
    Compatible with
    Size
    Ø int.
  • FKM
    6,1 mm
    CRS ONE O-rings
    CRS Inlet liner seals
    Agilent 8890, 7890, 6890 and 6850
    Thermo Scientific TRACE 1610, 1600, and 1300 series GC
    B010
    9,3 mm
    1 * 10 Pce
    1 * 10 Pce - 23,70 €
    23,70 €
    1 * 10 Pce
       23,70 €
  •  
    Material
    Ø
    Description
    For
    Compatible with
    Size
    Ø int.
  • FKM
    5,3 mm
    CRS ONE O-rings
    CRS Inlet liner seals
    Perkin Elmer Clarus 590 and 690
    B009
    8,8 mm
    1 * 10 ST
    1 * 10 ST - 23,70 €
    23,70 €
    1 * 10 ST
       23,70 €
  •  
    Material
    Ø
    Description
    For
    Compatible with
    Size
    Ø int.
  • FKM
    4,8 mm
    CRS ONE O-rings
    CRS Inlet liner seals
    Shimadzu Nexis GC-2030, GC-2014 and
    GC-2010
    P005
    8,6 mm
    1 * 10 ST
    1 * 10 ST - 23,70 €
    23,70 €
    1 * 10 ST
       23,70 €