Order Entry
Belgium
Orders LinkContactUsLinkComponent
L(+)-Ascorbic acid ≥99.0% USP

L(+)-Ascorbic acid ≥99.0% USP

Glass, amber, 25 g
Catalog # SPCMAS102-25GM
Supplier:   Spectrum Chemical
L(+)-Ascorbic acid ≥99.0% USP
Catalog # SPCMAS102-25GM
Supplier:  Spectrum Chemical
CAS Number:  50-81-7
L(+)-Ascorbic acid ≥99.0% USP
L(+)-Ascorbic acid ≥99.0% USP

Glass, amber, 25 g

CAS Number:  50-81-7
Price
81,50 €
Log in for availability
1 * 25 g
Product Details & Documents

Spectrum-labeled kosher products with the Kosher Supervision of America symbol are a firm guarantee to all Spectrum customers that the products bearing the symbol are in full compliance with the most demanding of Kosher and Pareve standards.

All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Synonyms:  Ascorbic Acid, L-Ascorbic acid, (R)-5-((S)-1,2-Dihydroxyethyl)-3,4-dihydroxyfuran-2(5H)-one, Acidum ascorbicum
  • CAS Number:50-81-7
  • Formula:C₆H₈O₆
  • MDL Number:MFCD00064328
  • Molecular Weight:176.13 g/mol
  • Storage Temperature:Ambient
Specifications

Specification Test Results

  • Assay:
    99.0 - 100.5 %
  • Specific Rotation [A]D:
    +20.5 to +21.5
  • Residue On Ignition:
    max. 0.1 %
  • Heavy Metals (As Pb):
    max. 20 ppm
  • Identification:
    To Pass Test

Specifications

  • Pack type:
    Glass, amber
  • Pk:
    25 g

Specifications

Aanbevelingen

Aanbevelingen worden alleen gepersonaliseerd op basis van uw winkelvoorkeuren als u daarvoor toestemming hebt gegeven door Verbeter mijn winkelervaring" op Mijn Account pagina" aan te zetten.

Anders ontvangt u algemene aanbevelingen.