ContactUsLinkComponent
Ultra-Low Temperature Freezers, TSX Universal
Ultra-Low Temperature Freezers, TSX Universal
Supplier:  Thermo Scientific
Certificates
undefined
Ultra-Low Temperature Freezers, TSX Universal
Supplier:  Thermo Scientific
Specifications
  • Catalog No:
  • 471-1315
  • 471-1316
  • 471-1317
  • 471-1318
  • Model:
  • TSX40086FA
  • TSX50086FA
  • TSX60086FA
  • TSX70086FA
  • Cat. No.:
  • HERATSX40086FA
  • HERATSX50086FA
  • HERATSX60086FA
  • HERATSX70086FA
  • Defrost:
  • Manual
  • Manual
  • Manual
  • Manual
  • Electrical:
  • 100 - 230 V, 50/60 Hz
  • 100 - 230 V, 50/60 Hz
  • 100 - 230 V, 50/60 Hz
  • 100 - 230 V, 50/60 Hz
  • Noise level:
  • <44 dBA
  • <44 dBA
  • <44 dBA
  • <44 dBA
  • Shelves/drawers:
  • 4 shelves (3 adjustable)
  • 4 shelves (3 adjustable)
  • 4 shelves (3 adjustable)
  • 4 shelves (3 adjustable)
  • Door type:
  • 1 outer, 2 inner doors
  • 1 outer, 2 inner doors
  • 1 outer, 2 inner doors
  • 1 outer, 2 inner doors
  • Weight:
  • 270 kg
  • 297 kg
  • 316 kg
  • 347 kg