Order Entry
Belgium
ContactUsLinkComponent
Epstein-Barr Virus (VCA) IgA ELISA kit
Epstein-Barr Virus (VCA) IgA ELISA kit
Catalog # ABNOKA6727.1KIT
Supplier:  Abnova
CAS Number:  
Epstein-Barr Virus (VCA) IgA ELISA kit
Catalog # ABNOKA6727.1KIT
Supplier:  Abnova
CAS Number:  

Specifications

  • Pk:
    1 KIT
  • Primary antibody reactivity:
    Epstein Barr virus
  • Target protein:
    IgA
  • Assay Principle:
    Qualitative

Specifications

About this item

Epstein-Barr Virus (VCA) IgA ELISA Kit is a qualitative determination of IgA class antibodies against Epstein-Barr Virus (VCA) in human serum or plasma.