Order Entry
Belgium
ContactUsLinkComponent
CCP Ab ELISA kit
CCP Ab ELISA kit
Catalog # ABNOKA6675.1KIT
Supplier:  Abnova
CAS Number:  
CCP Ab ELISA kit
Catalog # ABNOKA6675.1KIT
Supplier:  Abnova
CAS Number:  

Specifications

  • Pk:
    1 KIT
  • Target protein:
    CCP Ab
  • Assay Principle:
    Quantitative

Specifications

About this item

CCP Ab ELISA Kit is an enzyme-linked immunosorbent assay for quantitative detection of IgG class autoantibodies against cyclic citrullinated peptides (CCP) in human serum or plasma