Order Entry
Belgium
ContactUsLinkComponent
Recombinant Rat OSM (from E. coli)
Recombinant Rat OSM (from E. coli)
Catalog # ABNOP8999.10UG
Supplier:  Abnova
CAS Number:  
Recombinant Rat OSM (from E. coli)
Catalog # ABNOP8999.10UG
Supplier:  Abnova
CAS Number:  

Specifications

  • Pk:
    10 µG
  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Rat
  • Gene ID:
    Q65Z15
  • Protein/peptide name:
    OSM

Specifications

About this item