0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Belgium
ContactUsLinkComponent
Enzymes
Recombinant Alkaline Phosphatase (from E. coli)
Recombinant Alkaline Phosphatase (from E. coli)
ABNOP9384.100UL
:
Abnova
:
Recombinant Alkaline Phosphatase (from E. coli)
ABNOP9384.100UL
:
Abnova
:
Pk:
100 µl
Enzyme type:
Recombinant
Source:
E. coli
Enzyme name:
Alkaline Phosphatase