Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Belgium
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Enzymes
Recombinant Vaccinia virus capping enzyme (from E. coli)
Recombinant Vaccinia virus capping enzyme (from E. coli)
ABNOP9374.200UL
:
Abnova
:
Recombinant Vaccinia virus capping enzyme (from E. coli)
ABNOP9374.200UL
:
Abnova
:
Pk:
200 µl
Enzyme type:
Recombinant
Source:
E. coli
Enzyme name:
Vaccinia virus capping enzyme