Order Entry
Belgium
ContactUsLinkComponent
Recombinant mRNA Cap 2'-O-methyltransferase (from E. coli)
Recombinant mRNA Cap 2'-O-methyltransferase (from E. coli)
Catalog # ABNOP9376.50UL
Supplier:  Abnova
CAS Number:  
Recombinant mRNA Cap 2'-O-methyltransferase (from E. coli)
Catalog # ABNOP9376.50UL
Supplier:  Abnova
CAS Number:  

Specifications

  • Pk:
    50 µl
  • Enzyme type:
    Recombinant
  • Source:
    E. coli
  • Enzyme name:
    mRNA Cap 2'-O-methyltransferase

Specifications

About this item