Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Belgium
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Enzymes
Recombinant mRNA Cap 2'-O-methyltransferase (from E. coli)
Recombinant mRNA Cap 2'-O-methyltransferase (from E. coli)
Recombinant mRNA Cap 2'-O-methyltransferase (from E. coli)
Catalog # ABNOP9378.1ML
Supplier:
Abnova
CAS Number:
Expand image
Recombinant mRNA Cap 2'-O-methyltransferase (from E. coli)
Catalog # ABNOP9378.1ML
Supplier:
Abnova
CAS Number:
Specifications
Pk:
1 mL
Enzyme type:
Recombinant
Source:
E. coli
Enzyme name:
mRNA Cap 2'-O-methyltransferase
Specifications
About this item
Product Details
Product Details