0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Belgium
ContactUsLinkComponent
Enzymes
Recombinant Poly(A) Polymerase (from E. coli)
Recombinant Poly(A) Polymerase (from E. coli)
ABNOP9381.1ML
:
Abnova
:
Recombinant Poly(A) Polymerase (from E. coli)
ABNOP9381.1ML
:
Abnova
:
Pk:
1 mL
Enzyme type:
Recombinant
Source:
E. coli
Enzyme name:
Poly(A) Polymerase