Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Belgium
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Enzymes
Staphylococcus aureus Staphylokinase
Staphylococcus aureus Staphylokinase
Staphylococcus aureus Staphylokinase
Catalog # ABNOP7189.100UG
Supplier:
Abnova
CAS Number:
Expand image
Staphylococcus aureus Staphylokinase
Catalog # ABNOP7189.100UG
Supplier:
Abnova
CAS Number:
Specifications
Pk:
100 µG
Species:
Staphylococcus aureus
Enzyme name:
Staphylokinase
Specifications
About this item
Product Details
Product Details