Anti-Aspartate Aminotransferase Rabbit Recombinant Antibody [clone: R05-6C1]
ABNORAB01584.100UL
: Abnova
- Antigen symbol:Aspartate Aminotransferase
- Clonality:Recombinant
- Clone:R05-6C1
- Host:Rabbit
- Reactivity:Human
- Immunogen:Recombinant rabbit monoclonal antibody raised against synthetic peptide of human Aspartate Aminotransferase.
- Pk:100 µl