Anti-CD226 Rabbit Recombinant Antibody [clone: R01-4C1]
ABNORAB01526.100UL
: Abnova
- Antigen symbol:CD226
- Clonality:Recombinant
- Clone:R01-4C1
- Host:Rabbit
- Reactivity:Mouse
- Immunogen:Recombinant rabbit monoclonal antibody raised against mouse CD226.
- Pk:100 µl