Order Entry
Belgium
ContactUsLinkComponent
Anti-CD134 Rabbit Recombinant Antibody [clone: R01-5C7]
Anti-CD134 Rabbit Recombinant Antibody [clone: R01-5C7]
Catalog # ABNORAB01528.100UL
Supplier:  Abnova
Anti-CD134 Rabbit Recombinant Antibody [clone: R01-5C7]
Catalog # ABNORAB01528.100UL
Supplier:  Abnova

Specifications

  • Antigen symbol:
    CD134
  • Clonality:
    Recombinant
  • Clone:
    R01-5C7
  • Host:
    Rabbit
  • Reactivity:
    Mouse
  • Immunogen:
    Recombinant rabbit monoclonal antibody raised against mouse CD134.
  • Pk:
    100 µl

Specifications

About this item