Order Entry
Belgium
ContactUsLinkComponent
Anti-Calretinin Rabbit Recombinant Antibody [clone: R01-2G7]
Anti-Calretinin Rabbit Recombinant Antibody [clone: R01-2G7]
Catalog # ABNORAB01544.100UL
Supplier:  Abnova
Anti-Calretinin Rabbit Recombinant Antibody [clone: R01-2G7]
Catalog # ABNORAB01544.100UL
Supplier:  Abnova

Specifications

  • Antigen symbol:
    Calretinin
  • Clonality:
    Recombinant
  • Clone:
    R01-2G7
  • Host:
    Rabbit
  • Reactivity:
    Human
  • Immunogen:
    Recombinant rabbit monoclonal antibody raised against synthetic peptide of human Calretinin.
  • Pk:
    100 µl

Specifications

About this item