Order Entry
Belgium
ContactUsLinkComponent
Anti-CDK2 Rabbit Recombinant Antibody [clone: R06-4C1]
Anti-CDK2 Rabbit Recombinant Antibody [clone: R06-4C1]
Catalog # ABNORAB01792.100UL
Supplier:  Abnova
Anti-CDK2 Rabbit Recombinant Antibody [clone: R06-4C1]
Catalog # ABNORAB01792.100UL
Supplier:  Abnova

Specifications

  • Antigen symbol:
    CDK2
  • Clonality:
    Recombinant
  • Clone:
    R06-4C1
  • Host:
    Rabbit
  • Reactivity:
    Human
  • Immunogen:
    Recombinant rabbit monoclonal antibody raised against human CDK2.
  • Pk:
    100 µl

Specifications

About this item